Learn More
DWK Life Sciences Wheaton™ Battery Powered Crimping Tool
Built for durability and ergonomics, helping laboratory professionals easily crimp high volumes of sample vials without significant discomfort or distress. The high-performance tool features an external power source for consistently crimped vials. The battery powered tool allows for mobility in the lab.
Manufacturer:  DWK Life Sciences W225720
 View more versions of this product
Catalog No. 02-915-006
  Check Availability
Add to cart
Includes: 1 Charging Cord, 1 Lithium Ion Rechargeable Battery, 1 Product Information/Setup Guide.
Description
Description
Wheaton battery powered crimpers and decappers fit standard laboratory vials and seals with aluminum or two part aluminum/steel caps. The electronically adjustable cycle is accurate and reproducible and can be adjusted with the control pad on top of the unit. The tools are ergonomically designed to reduce strain and arm injury associated with many manual crimping tools.
Rechargable: The tools can be recharged in one to two hours with the universal power supply included. The power supply includes a plug set for operation in most countries
  • Powerful motor with a fast cycle
  • Convenient controls on top of unit to adjust crimp
  • Advanced charging system including warning light when recharging is necessary
  • Long lasting battery with Lithium Ion technology – up to 800 crimps per charge
  • Advanced charging system including warning light when recharging is necessary
  • Charges in 1 to 2 hours
  • Can be operated while charging
  • Optional base and mounting kit (sold separately)
Specifications
Specifications
Battery Powered Crimping Tool
20mm
Crimping autosampler vials
1 charging cord, 1 Lithium Ion rechargeable battery, 1 Product Information/Setup Guide