Learn More
Bausch & Lomb™ Hastings Triplet Adjustable Focus Measuring Magnifying Glass
Provides a flat image without distortion
Manufacturer:  BAUSCH & LOMB 813433
 View more versions of this product
Catalog No. 12-032-03
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
40 diopter
Protective case and storage pouch for case and lens
2.54cm
10x
To provide a flat image without distortion
1 in.
Glass
Hastings Triplet Measuring Magnifier