Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bausch & Lomb™ Hastings Triplet Magnifiers

Three separate lenses bonded together to form a compound lens

Manufacturer:  BAUSCH & LOMB 816168

 View more versions of this product

Catalog No. 12-032A


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

0.78 in.
2cm
Used by professionals
1.5 in.
Glass
Hastings Triplet Magnifier
1.98cm
28 diopter
Protective pouch for storage
3.81cm
7x
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit