Bausch & Lomb™ Scales for Magnifiers
Designed for use with Lenscope Magnifiers and Hasting Triplet Measuring Magnifiers
Manufacturer: BAUSCH & LOMB 813437
 View more versions of this product
Catalog No. 17-985-558
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) To measure 0.75 in. in 0.005 in. intervals
    For Use With (Equipment) Lenscope™ magnifier
    Includes Protective storage pouch
    Material Glass
    Product Type Inch Scale
  • Description & Specifications
Related Products
Product Content Feedback