Learn More
Bax Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and flow cytometry experiments.
Supplier: Epredia MS711R1
Description
Bcl-2, Bax, and Bcl-X are members of a family of proteins that regulate apoptosis. In contrast to Bcl-2α and Bcl-XL, overexpression of bax protein accelerates the programmed cell death (apoptosis).Host Species: Mouse
Clone: 2D2
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with mouse and rat. Others not tested.
Epitope: aa 3-16
Immunogen: A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human Bax protein. This amino acid sequence is not shared by mouse and rat Bax protein.
Molecular Weight: 21kDa
Positive Control: HeLa cells, Hodgkin's lymphoma
Cellular Localization: Cytoplasmic and cell membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Flow Cytometry (Not tested)
Note:
USA: RUO; Int'l: RUO
Specifications
Bax Ab-1 | |
Monoclonal | |
Unconjugated | |
Mouse | |
1 mL | |
Apoptosis | |
Human | |
IgG2a κ |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2D2 | |
Ready-to-use for IHC (UVLP) | |
A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human bax protein. This amino acid sequence is NOT shared by mouse and rat bax protein. | |
RUO | |
Primary | |
Prediluted |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.