Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Bax Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry,  western blotting, immunofluorescence, and flow cytometry experiments.

$171.10 - $966.50

Specifications

Antigen Bax Ab-1
Clone 2D2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 10
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS711B
View Documents
Thermo Scientific™
MS711B
Biotinylated Ab with BSA and Azide 1mL Each for $937.00
Add to cart
 
MS711B0
View Documents
Thermo Scientific™
MS711B0
Biotinylated Ab with BSA and Azide 100μL Each for $228.00
Add to cart
 
MS711B1
View Documents
Thermo Scientific™
MS711B1
Biotinylated Ab with BSA and Azide 500μL Each for $621.50
Add to cart
 
MS711P
View Documents
Thermo Scientific™
MS711P
Purified Ab with BSA and Azide 1mL Each for $728.50
Add to cart
 
MS711P0
View Documents
Thermo Scientific™
MS711P0
Purified Ab with BSA and Azide 100μL Each for $171.10
Add to cart
 
MS711P1
View Documents
Thermo Scientific™
MS711P1
Purified Ab with BSA and Azide 500μL Each for $478.50
Add to cart
 
MS711P1ABX
View Documents
Thermo Scientific™
MS711P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $531.50
Add to cart
 
MS711PABX
View Documents
Thermo Scientific™
MS711PABX
Ab without BSA/Azide (Frozen) 200μL Each for $966.50
Add to cart
 
MS711R7
View Documents
Thermo Scientific™
MS711R7
Ready-to-use for IHC (UVLP) 7mL Each for $413.00
Add to cart
 
MS711R1
View Documents
Thermo Scientific™
MS711R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, and flow cytometry experiments with Thermo Scientific™ Bax Ab-1, Mouse Monoclonal Antibody.

Bcl-2, Bax, and Bcl-X are members of a family of proteins that regulate apoptosis. In contrast to Bcl-2α and Bcl-XL, overexpression of bax protein accelerates the programmed cell death (apoptosis).

Host Species: Mouse

Clone: 2D2

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and rat. Others not tested.

Epitope: aa 3-16

Immunogen: A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human Bax protein. This amino acid sequence is not shared by mouse and rat Bax protein.

Molecular Weight: 21kDa

Positive Control: HeLa cells, Hodgkin's lymphoma

Cellular Localization: Cytoplasmic and cell membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry (Not tested)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Bax Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Murine
Apoptosis
A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human bax protein. This amino acid sequence is NOT shared by mouse and rat bax protein.
2D2
Monoclonal
RUO
Primary
IgG2a kappa
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit