Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Bax Ab-4, Mouse Monoclonal Antibody

Thermo Scientific™ Bax Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

Manufacturer:  Thermo Scientific™ MS714P1

 View more versions of this product

Catalog No. MS714P1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Thermo Scientific™ Bax Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results. 

Bcl-2, Bax, and Bcl-X are members of a family of proteins that regulate apoptosis. In contrast to Bcl-2α and Bcl-XL, overexpression of bax protein accelerates the programmed cell death (apoptosis).

Host Species: Mouse

Clone: 6A7

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: aa 12-24

Immunogen: A Synthetic peptide, aa 12-24 (Cys-GPTSSEQIMKTGA), of human Bax protein. This amino acid sequence is shared by human, mouse and rat Bax protein.

Molecular Weight: 21kDa

Positive Control: PC12 or HeLa cells

Cellular Localization: Cytoplasmic and cell membrane

Recommended for:

  • Western Blotting (Not optimum)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Bax Ab-4
Monoclonal
200μg/mL
Concentrated
A Synthetic peptide, aa 12-24 (Cys-GPTSSEQIMKTGA), of human Bax protein. This amino acid sequence is shared by human, mouse and rat Bax protein.
500μL
Apoptosis
Primary
Human, Murine, Rat
Immunofluorescence
6A7
Unconjugated
Murine
IgG1
RUO
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit