Learn More
Invitrogen™ Bax Monoclonal Antibody (2D2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA513994
Description
MA5-13994 targets Bax in IF, IHC (P), and WB applications and shows reactivity with Human samples. The MA5-13994 immunogen is a synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human bax protein. This amino acid sequence is NOT shared by mouse and rat bax protein.
BAX is a members of the Bcl-2 Family and plays an important role in regulation of apoptosis. Whereas Bcl-2 is commonly regarded as an anti-apoptotic protein, BAX is considered to have a pro-apoptotic function. Regulation of apoptosis is supposed to involve both homo- and heterodimerization of different isoforms of BAX and Bcl-2. The Bax gene encodes different isoforms including Bax alpha (21 kDa) and Bax beta (24 kDa), whereas both isoforms contain the BH1, BH2 and BH3 domains, Bax beta has a unique carboxyl terminus and does not contain a hydrophobic transmembrane domain. Bcl-2 is also expressed in different Isoforms. Bcl-2 beta differs in the 3' UTR and coding region compared to variant alpha. Bcl-2 beta is shorter (22 kDa) and has a distinct C-terminus compared to Bcl-2 alpha (26 kDa). BAX is a member of the BCL-2 family of proteins, which function as regulators of apoptosis. Overexpression of BAX functions to promote cell death. BAX can form homodimers and is also able to heterodimerize with other BCL-2 related proteins.
Specifications
Bax | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
Q07812 | |
BAX | |
A synthetic peptide, aa 3-16 (Cys-GSGEQPRGGGPTSS) of human bax protein. This amino acid sequence is NOT shared by mouse and rat bax protein. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin), Western Blot | |
2D2 | |
Unconjugated | |
BAX | |
apoptosis regulator BAX; Apoptosis regulator Bcl-X; Bax; Bax zeta; BAX-ALPHA; Bax-alpha protein; Baxdelta2G9; Baxdelta2G9omega; Baxdelta2omega; BCL2 associated X protein; BCL2 associated X, apoptosis regulator; BCL2 like 1; BCL2-associated protein; Bcl-2-associated protein Bax; Bcl2-associated protein Bax; Bcl-2-associated X protein; BCL2-associated X protein; BCL2-associated X protein omega; BCL2L; BCL2L1; Bcl2-L-1; BCL2L4; Bcl2-L-4; BCL2-like 1; bcl-2-like protein 1; Bcl-2-like protein 4; BCLX; Bcl-X; BCL-XL/S; PPP1R52; protein phosphatase 1, regulatory subunit 52 | |
Mouse | |
Protein G | |
RUO | |
581 | |
4°C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.