Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAX Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5120126

This item is not returnable.
View return policy
Description
Positive test controls include: HeLa, MCF-7, HT-1080. The target is usually found in the following locations: Cytoplasm, Cytoplasm, Mitochondrion membrane, Single-pass membrane protein.
BAX Polyclonal antibody specifically detects BAX in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western BlotSpecifications
BAX | |
Polyclonal | |
Unconjugated | |
BAX | |
apoptosis regulator BAX; Apoptosis regulator Bcl-X; Bax; Bax zeta; BAX-ALPHA; Bax-alpha protein; Baxdelta2G9; Baxdelta2G9omega; Baxdelta2omega; BCL2 associated X protein; BCL2 associated X, apoptosis regulator; BCL2 like 1; BCL2-associated protein; Bcl-2-associated protein Bax; Bcl2-associated protein Bax; Bcl-2-associated X protein; BCL2-associated X protein; BCL2-associated X protein omega; BCL2L; BCL2L1; Bcl2-L-1; BCL2L4; Bcl2-L-4; BCL2-like 1; bcl-2-like protein 1; Bcl-2-like protein 4; BCLX; Bcl-X; BCL-XL/S; PPP1R52; protein phosphatase 1, regulatory subunit 52 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human Bax (NP_620116.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
2.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q07812, Q07813, Q63690 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12028, 24887, 581 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title