Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bax Rabbit anti-Human, KHL, Clone: 1C7, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen Bax
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB110325MI
View Documents
MilliporeSigma
ZRB110325UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB11034XMI
View Documents
MilliporeSigma
ZRB11034X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Bax Monoclonal antibody specifically detects Bax in Human samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

Bax
KHL
Rabbit
Apoptosis regulator BAX;Bcl-2-like protein 4;Bcl2-L-4
KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal region of human Apoptosis regulator BAX.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Purified
Human
BAX, BCL2L4
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit