Bay 59-3074, Tocris

CB1/CB2 receptor partial agonist

Manufacturer: R&D Systems 2500/50

Quantity 50mg

 View more versions of this product

Catalog No. 25-005-0

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C18H13F6NO4S
    Formula Weight 453.35
    Synonym 3-[2-Cyano-3-(trifluoromethyl)phenoxy]phenyl 4, 4, 4-trifluoro-1-butanesulfonic acid ester
    Chemical Name or Material Bay 59-3074

    Specifications

    Formula Weight 453.35
    Quantity 50mg
    Assay Percent Range >99%
    Molecular Formula C18H13F6NO4S
    CAS 406205-74-1
    Synonym 3-[2-Cyano-3-(trifluoromethyl)phenoxy]phenyl 4, 4, 4-trifluoro-1-butanesulfonic acid ester
    Recommended Storage Store at +4°C
    Chemical Name or Material Bay 59-3074

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit