Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAY 60-6583, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent adenosine A2B receptor agonist (EC50 = 2.83 nM for murine A2B receptor). Displays selectivity for A2B over A1, A2A and A3 receptors. Decreases fMLP-induced superoxide production in neutrophils at low concentrations (1-10 nM). Cardioprotective; attenuates infarct size in a mouse model of myocardial ischemia. Exhibits ERK1/2-biased agonism.
Specifications
Quantity | 10 mg |
Formula Weight | 379.44 |
Percent Purity | >98% |
Chemical Name or Material | BAY 60-6583 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction