Learn More
Medchemexpress LLC BAY-728 | C24H28F3N5O2S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY153045A10MM/1ML
Encompass_Preferred
BAY-728 serves as a negative control for BAY-805, which is described as a potent and selective inhibitor of USP21. This product is intended for research use only and is not sold to patients. It appears as a white to off-white solid with a molecular weight of 507.57.
  • Used as a negative control for BAY-805
  • Potent and selective inhibitor of USP21
  • Intended for research use only
  • Appears as a white to off-white solid
  • Suitable for laboratory chemicals and manufacturing substances
Catalog No. 50-003-72039