Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (±)-Bay K 8644

Click to view available options
Quantity:
5mg
71145-03-4
C16H15F3N2O4
356.301
MFCD00036697
ZFLWDHHVRRZMEI-UHFFFAOYSA-N
methyl 2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-1,4-dihydropyridine-3-carboxylate,3-pyridinecarboxylic acid, 1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-, methyl ester,methyl 2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-1,4-dihydropyridine-3-carboxylate,1,4-dihydro-2,6-dimethyl-5-nitro-4-2′-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester,3-pyridinecarboxylic acid, 1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-, methyl ester,bay-k-8644, +--isomer,opera_id_789,d01rad,y-bay k 8644,r-+-1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester
2303
CHEBI:34555
methyl 2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-1,4-dihydropyridine-3-carboxylate
CC1=C(C(C(=C(N1)C)[N+](=O)[O-])C2=CC=CC=C2C(F)(F)F)C(=O)OC
71145-03-4
C16H15F3N2O4
356.301
MFCD00036697
ZFLWDHHVRRZMEI-UHFFFAOYSA-N
methyl 2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-1,4-dihydropyridine-3-carboxylate,3-pyridinecarboxylic acid, 1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-, methyl ester,methyl 2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-1,4-dihydropyridine-3-carboxylate,1,4-dihydro-2,6-dimethyl-5-nitro-4-2′-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester,3-pyridinecarboxylic acid, 1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-, methyl ester,bay-k-8644, +--isomer,opera_id_789,d01rad,y-bay k 8644,r-+-1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester
2303
CHEBI:34555
methyl 2,6-dimethyl-5-nitro-4-[2-(trifluoromethyl)phenyl]-1,4-dihydropyridine-3-carboxylate
CC1=C(C(C(=C(N1)C)[N+](=O)[O-])C2=CC=CC=C2C(F)(F)F)C(=O)OC

An active L-type, voltage-gated calcium channel agonist

Color Yellow
Quantity 5mg
Solubility Information Soluble in DMSO, ethanol, or methanol; Insoluble in water
Formula Weight 356.3
Physical Form Powder
Chemical Name or Material (±)-Bay K 8644

RTECSNumber : US5655000