Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bay Oil, FCC, 50-65%, Spectrum™

Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Amber Glass Bottle
8006-78-8
8006-78-8

B1061, 8006-78-8

Spectrum™ Bay Oil, FCC is used in cooking as a flavoring agent. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Refractive Index 1.507-1.516
Quantity 25 mL
CAS Max % 1
UN Number UN1197
Percent Purity 50 to 65%