Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers Bayer™ Model 600 Series Electrolyte Analyzers: Electrodes

Electrodes for use with Bayer™ 600 Series Electrolyte Analyzers.

Manufacturer:  Siemens Healthineers 476267000

 View more versions of this product

Catalog No. 09-529-252


Add to cart

Description

Description

For use with Bayer Electrolyte Analyzer models 614 and 634.
Specifications

Specifications

pH Electrode2
Electrolyte analysis
Bayer Electrolyte Analyzers: 614, 634
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit