Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAZ2A Polyclonal Antibody, Active Motif™
SDP

Rabbit Polyclonal Antibody

$145.44 - $572.16

Specifications

Antigen BAZ2A
Concentration Lot-Specific
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3641
SDP
Encompass
View Documents
Active Motif
61708
10 μL
Each for $145.44
Add to Cart
 
50-199-3640
SDP
Encompass
View Documents
Active Motif
61707
100 μL
Each for $572.16
Add to Cart
 
Description

Description

TIP 5 is the large subunit of the nucleolar remodeling complex NoRC. NoRC causes the repression of ribosomal gene transcription. It was demonstrated that histone deacetylation is involved in this repression and that TIP5 is associated with the histone deacetylase HDAC1 and mediates the deacetylation of histones in the vicinity of the rDNA promoter. The interaction of TIP5 and HDAC1, which is necessary for transcriptional repression, is established by the C-terminal PHD finger and bromodomain.
Specifications

Specifications

BAZ2A
Western Blot
Unconjugated
Rabbit
Human
Q9UIF9
11176
This antibody was raised against a peptide within the N-terminal region of human BAZ2A.
Primary
-20°C, Avoid Freeze/Thaw Cycles
BAZ2A
Lot-Specific
Polyclonal
Liquid
RUO
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
AA415431; BAZ2A; bromodomain adjacent to zinc finger domain 2A; Bromodomain adjacent to zinc finger domain protein 2A; bromodomain adjacent to zinc finger domain, 2A; C030005G16Rik; C78388; hWALp3; KIAA0314; mKIAA0314; TIP5; Transcription termination factor I-interacting protein 5; TTF-I interacting peptide 5; TTF-I-interacting protein 5; Walp3
BAZ2A
IgG
Antigen affinity chromatography
Antibody
SDS
Documents

Documents

Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit