Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ Bazedoxifene acetate

Potent and selective estrogen receptor modulator (SERM)

Manufacturer:  Tocris™ 5263/10

 View more versions of this product

Catalog No. 52-631-0


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Potent selective estrogen receptor modulator (SERM) (IC50 values are 26 and 99nM for ERα and ERβ respectively). Inhibits 17β-estradiol-induced proliferation in MCF-7 cells. Co-treatment with Raloxifene (Cat No. 2280) completely abolishes raloxifene-induced stimulation of luminal epithelial cells and myometrium.
  • Chemical Name: 1-[[4-[2-(Hexahydro-1H-azepin-1-yl)ethoxy]phenyl]methyl]-2-(4-hydroxyphenyl)-3-methyl-1H-indol-5-ol acetate
  • Formula: C30H34N2O3.C2H4O2
  • CAS Number: 198481-33-3
  • Solubility: Soluble to 100mM in DMSO and to 20mM in ethanol
  • Purity: >98
Description & Specifications

Specifications

Bazedoxifene acetate
>98
10mg
Bazedoxifene acetate
530.65
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit