Learn More
BBOX1 Mouse anti-Human, Clone: OTI3B6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505752

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
BBOX1 encodes gamma butyrobetaine hydroxylase which catalyzes the formation of L-carnitine from gamma-butyrobetaine, the last step in the L-carnitine biosynthetic pathway. Carnitine is essential for the transport of activated fatty acids across the mitochondrial membrane during mitochondrial beta-oxidation.Specifications
BBOX1 | |
Monoclonal | |
Unconjugated | |
BBOX1 | |
BBH, BBOX, G-BBH, gamma-BBH | |
Mouse | |
Affinity Chromatography | |
RUO | |
8424 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3B6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O75936 | |
BBOX1 | |
Full length human recombit protein of human BBOX1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.