Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BBS1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$204.00 - $482.50

Specifications

Antigen BBS1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP17968420
SDP
View Documents
Novus Biologicals
NBP17968420UL
20 μL
Each for $204.00
Add to Cart
 
NBP179684
SDP
View Documents
Novus Biologicals
NBP179684
100 μL
Each for $482.50
Add to Cart
 
Description

Description

BBS1 Polyclonal specifically detects BBS1 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

BBS1
Polyclonal
Rabbit
NP_001028300
582
Synthetic peptide directed towards the N terminal of human Bbs1The immunogen for this antibody is Bbs1. Peptide sequence PCVYVYKNLRPYFKFSLPQLPPNPLEQDVWNQAKEDQIDPLTLKEMLEDI.
Primary
Western Blot
Unconjugated
RUO
Bardet-Biedl syndrome 1, Bardet-Biedl syndrome 1 protein, BBS2L2, BBS2-like protein 2, FLJ23590, MGC126183, MGC126184, MGC51114
BBS1
IgG
65 kDa
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit