Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BBS4 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI2D5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270241AF647

Catalog No. N270241A647


Add to cart

Description

Description

BBS4 Monoclonal antibody specifically detects BBS4 in Human samples. It is validated for Western Blot.
Specifications

Specifications

BBS4
OTI2D5
Western Blot
Affinity Purified, Conjugated
BBS4
Recombinant protein fragment corresponding to amino acids 266-519 of human BBS4 (NP_149017) produced in E.coli.
Immunogen affinity purified
Stem Cell Markers, Vision
Primary
585.0
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Bardet-Biedl syndrome 4, Bardet-Biedl syndrome 4 protein
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit