Learn More
Aviva Systems Biology Corp BBS5 Antibody - middle region (ARP52931_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52931_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BBS5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BBS5. Alternate names: -ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-60872