Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BC 11-38, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
686770-80-9
C15H16N2OS2
304.426
YHNDCCKFNWDQGW-UHFFFAOYSA-N
d03qth,3-phenyl-2-propylsulfanyl-6,7-dihydrothieno 3,2-d pyrimidin-4-one,3-phenyl-2-propylsulfanyl-3h,4h,6h,7h-thieno 3,2-d pyrimidin-4-one,3-phenyl-2-propylsulfanyl-6h,7h-thieno 3,2-d pyrimidin-4-one,3-phenyl-2-propylthio-6,7-dihydrothieno 3,2-d pyrimidin-4 3h-one,6,7-dihydro-3-phenyl-2-propylthio thieno 3,2-d pyrimidin-4 3h one
2052828
3-phenyl-2-propylsulfanyl-6,7-dihydrothieno[3,2-d]pyrimidin-4-one
CCCSC1=NC2=C(C(=O)N1C3=CC=CC=C3)SCC2
This item is not returnable. View return policy
686770-80-9
C15H16N2OS2
304.426
YHNDCCKFNWDQGW-UHFFFAOYSA-N
d03qth,3-phenyl-2-propylsulfanyl-6,7-dihydrothieno 3,2-d pyrimidin-4-one,3-phenyl-2-propylsulfanyl-3h,4h,6h,7h-thieno 3,2-d pyrimidin-4-one,3-phenyl-2-propylsulfanyl-6h,7h-thieno 3,2-d pyrimidin-4-one,3-phenyl-2-propylthio-6,7-dihydrothieno 3,2-d pyrimidin-4 3h-one,6,7-dihydro-3-phenyl-2-propylthio thieno 3,2-d pyrimidin-4 3h one
2052828
3-phenyl-2-propylsulfanyl-6,7-dihydrothieno[3,2-d]pyrimidin-4-one
CCCSC1=NC2=C(C(=O)N1C3=CC=CC=C3)SCC2

Selective PDE11 inhibitor

Selective phosphodiesterase (PDE) 11 inhibitor (IC50 = 0.28 μM for PDE11; IC50 values are >100 μM for PDE1 - PDE10). Significantly elevates cAMP levels and cortisol production in H295R human adenocarcinoma cells.
Quantity 50mg
Assay Percent Range >98%
Formula Weight 304.43
Chemical Name or Material BC 11-38

Recommended Storage : Store at +4°C