Learn More
BioVision IncSupplier Diversity Partner BCA Protein Assay Kit - Reducing Agent Compatible: Only kit on the market that is compatible with higher conc. of reducing agents TCEP (20 mM), DTT (10 mM), β-ME (35 mM) DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K8181000
BCA Protein Assay Kit - Reducing Agent Compatible: BioVision's Reducing Agent Compatible BCA Protein Assay Kit is the only protein estimation kit on the market that is compatible with strong reducing agents such as TCEP up to 20 mM, DTT up to 10 mM & ?-mercaptoethanol up to 35 mM. This kit is adapted from BioVision's BCA Assay Kit II and is based on the chelation of bicinchoninic acid (BCA) with the cuprous cation (Cu+1), which is generated by reduction of cupric cation (Cu+2) with the protein in alkaline conditions. BioVision's Reducing Agent Compatible BCA Protein Assay Kit is useful for determing protein concentrations of samples containing DTT, TCEP & ?-mercaptoethanol. The assay is linear over a wide range of protein concentration between 25-2000 ?g/ml. In general, protein concentrations are calculated with reference to a commonly used protein standard. The kit also includes Bovine Serum Albumin (BSA) as a protein standard for estimation of total protein content of samples.
Catalog No. 50-111-9933