Learn More
BioVision IncSupplier Diversity Partner BCA Protein Quantitation Kit: BCA Protein Assay Kit: Simple, Rapid & Detergent tolerant (up to 5 %) Assay for Determining Protein concentration in solution. Detection Range: 0.5 – 30 μg (0.01 - 0.6 mg/ml). 1000 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K8121000
The BCA Protein Assay Kit provides a simple, rapid, detergent tolerant (up to 5%) procedure for determining the concentration of proteins in solution. The method utilizes a copper (Cu²ᶧ) salt which can be reduced to the cuprous state by protein(s). The generated Cu²ᶧ ion forms an intensely colored complex with the bicinchoninic acid reagent with a very strong absorbance band centered at 562 nm. The intensity of the blue complex is proportional to the amount of protein in the sample. The BCA Protein Assay is suitable for measuring protein concentration in the range of 0.5-30 μg protein (0.01-0.6 mg/ml). BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9928