Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAM Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550376

Catalog No. PA550376


Add to cart

Description

Description

The antibody detects endogenous levels of total BCAM / CD239 protein.

Lutheran blood group glycoprotein is a member of the immunoglobulin superfamily and a receptor for the extracellular matrix protein, laminin. The protein contains five, N-terminus, extracellular immunoglobulin domains, a single transmembrane domain, and a short, C-terminal cytoplasmic tail. This protein may play a role in epithelial cell cancer and in vaso-occlusion of red blood cells in sickle cell disease. Two transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

BCAM
Polyclonal
Unconjugated
BCAM
Liquid
BCAM
Fusion protein corresponding to a region derived from internal residues of human basal cell adhesion molecule (Lutheran blood group)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1.9 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
P50895, Q9R069
AU, LU, CD239, MSK19
Rabbit
IgG
100 μL
-20°C
Primary
4059, 57278
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit