Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCAM/CD239 Rabbit anti-Human, Biotin, Clone: 2, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289356B
Description
BCAM/CD239 Monoclonal specifically detects BCAM/CD239 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
BCAM/CD239 | |
Monoclonal | |
Biotin | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
4059 | |
Human | |
Purified |
ELISA | |
2 | |
ELISA, Sandwich ELISA Detection | |
antigen identified by monoclonal F8, Auberger B antigen, basal cell adhesion molecule, basal cell adhesion molecule (Lu and Au blood groups), basal cell adhesion molecule (Lutheran blood group), B-CAM cell surface glycoprotein, B-cell adhesion molecule, CD239, CD239 antigen, F8/G253 antigen, glycoprotein 95kDa, LUAU, Lutheran antigen, Lutheran blood group (Auberger b antigen included), Lutheran blood group glycoprotein, MSK19 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human BCAM/CD239 (NP_005572.2; Met1-Ala547). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Human ALCAM Human CD31 Human EpCAM Human ICAM1 Human ICAM2 Human ICAM3 Human MCAM Human VCAM1 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title