Learn More
PIK3AP1 Mouse anti-Human, Non-human primate, Rat, Clone: OTI6H6, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA525250
Catalog No. PIMA525250
Add to cart
Description
Description
Signaling adapter that contributes to B-cell development by linking B-cell receptor (BCR) signaling to the phosphoinositide 3-kinase (PI3K)-Akt signaling pathway. Has a complementary role to the BCR coreceptor CD19, coupling BCR and PI3K activation by providing a docking site for the PI3K subunit PIK3R1. Alternatively, links Toll-like receptor (TLR) signaling to PI3K activation, a process preventing excessive inflammatory cytokine production. Also involved in the activation of PI3K in natural killer cells. May be involved in the survival of mature B-cells via activation of REL.
Specifications
Specifications
PIK3AP1
Monoclonal
0.85 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q6ZUJ8
Pik3ap1, 1810044J04Rik, B cell adaptor protein, BCAPB-cell adapter for phosphoinositide 3-kinase, B-cell phosphoinositide 3-kinase adapter protein 1, FLJ35564, phosphoinositide 3-kinase adapter protein 1, phosphoinositide-3-kinase adaptor protein 1, RP11-34E5.3
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
118788, 294048, 705017
Flow Cytometry, Western Blot
OTI6H6
Unconjugated
PIK3AP1
Liquid
PIK3AP1
Full length human recombit protein of PIK3AP1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Non-human primate, Rat