Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAP31 Rat anti-Human, Clone: CC-1, Abnova™

Rat monoclonal antibody raised against human BCAP31.

Manufacturer:  Abnova Corporation MAB12541

Catalog No. 89-933-624


Add to cart

Description

Description

This gene encodes a member of the B-cell receptor associated protein 31 superfamily. The encoded protein is a multi-pass transmembrane protein of the endoplasmic reticulum that is involved in the anterograde transport of membrane proteins from the endoplasmic reticulum to the Golgi and in the caspase 8-mediated apoptosis. Microdeletions in this gene are associated with the contiguous ABCD1/DXS1375E deletion syndrome. Two pseudogenes have been identified on chromosome 16. Alternatively spliced transcript variants encoding distinct isoforms have been described although the biological validity of some of the variants has not been determined. [provided by RefSeq]

Specifications

Specifications

BCAP31
CC-1
Rat monoclonal antibody raised against human BCAP31.
In PBS, pH 7.2 (0.02% sodium azide).
6C6-AG/BAP31/CDM/DXS1357E
Rat
IgG2a
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA Immunocytochemistry (1:1000-1:10000) Immunohistochemistry (Frozen sections) (1:1000-1:10000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5 ug/mL) Immunoprecipitation Western Blot (1:1000-1:6000) The opti
Liquid
BCAP31
Recombinant fusion protein corresponding to amino acids 230-246 of human BCAP31.
Immunoaffinity purification
RUO
Primary
10134
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit