Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551116

Catalog No. PA551116


Add to cart

Description

Description

BCAT1 Polyclonal Antibody detects endogenous levels of BCAT1 protein.

This gene encodes the cytosolic form of the enzyme branched-chain amino acid transaminase. This enzyme catalyzes the reversible transamination of branched-chain alpha-keto acids to branched-chain L-amino acids essential for cell growth. Two different clinical disorders have been attributed to a defect of branched-chain amino acid transamination: hypervalinemia and hyperleucine-isoleucinemia. As there is also a gene encoding a mitochondrial form of this enzyme, mutations in either gene may contribute to these disorders. Alternatively spliced transcript variants have been described.
Specifications

Specifications

BCAT1
Polyclonal
Unconjugated
BCAT1
Liquid
BCAT1
Synthesized peptide derived from the Internal region of human ECA39.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P54687, P24288, P54690
BCAT1, BCT1, ECA39, Branched-chain-amino-acid aminotransferase, cytosolic, BCAT(c), Protein ECA39
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
12035, 29592, 586
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit