Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCAT2 Antibody (7G3A11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261699
Description
BCAT2 Monoclonal specifically detects BCAT2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
BCAT2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
BCAM, BCAT(m), BCATM, BCT2EC 2.6.1.42, branched chain amino-acid transaminase 2, mitochondrial, branched chain aminotransferase 2, mitochondrial, branched-chain-amino-acid aminotransferase, mitochondrial, ECA40, Placental protein 18, PP18 | |
Mouse | |
44.3 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
7G3A11 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
BCAT2 | |
Purified recombinant fragment of human BCAT2 (AA: 259-393) expressed in E. Coli. | |
Protein G purified | |
metabolism | |
587 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction