Learn More
Seracare Life Sciences IncSupplier Diversity Partner BCIP/NBT WESTERN BLOTTING KIT  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 54100013
The Protein Detector Western Blot Kit is designed for the colorimetric detection of proteins immobilized on membranes through either electrophoresis or dot blotting. The combination of highly specific mouse and rabbit secondary antibody-alkaline phosphatase conjugates with a sensitive chromogenic substrate allows rapid and accurate identification of samples. All solutions required for blocking and washing the membrane and for diluting antibodies are provided. For added flexibility, affinity purified anti-mouse and anti-rabbit secondary antibodies conjugated with alkaline phosphatase are provided in stabilized, liquid form for quick dilution. The BCIP/NBT substrate is provided as a convenient, ready-to-use solution, which forms an insoluble purple precipitate when reacted with the AP-labeled antibody. The kit is ideal for routine colorimetric detection of overexpressed protein; sensitivity level to 500 pg.
Catalog No. 50-675-19