Learn More
BCKDK Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA549761
Catalog No. PA549761
Add to cart
Description
Description
The antibody detects endogenous levels of total BCKDK protein.
The second major step in the catabolism of the branched-chain amino acids, isoleucine, leucine, and valine, is irreversibly catalyzed by the branched-chain alpha-keto acid dehydrogenase complex (BCKD), an inner-mitochondrial enzyme complex composed of 3 catalytic components: a branched-chain alpha-keto acid decarboxylase (E1), a dihydrolipoyl transacylase (E2), and a dihydrolipoamide dehydrogenase (E3). The complex also contains 2 enzymes that regulated the state of activity of the BCKD complex: a kinase (BCKDK), and a phosphorylase. The ubiquitiously expressed kinase contains 1 histidine kinase domain. Maple syrup urine disease (MSUD) is a pathology secondary to an enzyme defect in the catabolic pathway of leucine, isoleucine, and valine. Accumulation of these amino acids and their corresponding keto acids results in encephalopathy and progressive neurodegeneration in infants not treated for MSUD.
Specifications
Specifications
BCKDK
Polyclonal
Unconjugated
BCKDK
Liquid
BCKDK
Synthesized peptide derived from Internal of human BCKD.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O14874, O55028, Q00972
[3-methyl-2-oxobutanoate dehydrogenase [lipoamide]] kinase, mitochondrial [Precursor], EC 2.7.11.4, Branched-chain alpha-ketoacid dehydrogenase kinase, BCKDHKIN
Rabbit
IgG
100 μL
-20°C
Primary
10295, 12041, 29603