Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-10 Mouse anti-Human, Clone: rBL10/411, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen Bcl-10
Clone rBL10/411
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122412
View Documents
Novus Biologicals
NBP30746920UG
20 μg
Each for $159.82
Add to cart
 
NB122411
View Documents
Novus Biologicals
NBP30746910UG
100 μg
Each for $374.34
Add to cart
 
Description

Description

Bcl-10 Monoclonal antibody specifically detects Bcl-10 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)
Specifications

Specifications

Bcl-10
0.2 mg/ml
Flow Cytometry, Immunohistochemistry (Paraffin)
Purified
RUO
Human
B-cell CLL/lymphoma 10CIPERCLAPB-cell lymphoma/leukemia 10, Bcl-10, CARD-containing apoptotic signaling protein, CARD-containing molecule enhancing NF-kappa-B, CARD-containing proapoptotic protein, CARD-like apoptotic protein, CARMEN, caspase-recruiting domain-containing protein, cCARMEN, c-E10bcl-10, CED-3/ICH-1 prodomain homologous E10-like regulator, Cellular homolog of vCARMEN, cellular-E10, hCLAP, Mammalian CARD-containing adapter molecule E10, mE10CARD containing molecule enhancing NF-kB
Human Bcl-10 recombinant protein (epitope aa122-168)
Monoclonal
Protein A or G purified
rBL10/411
Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Apoptosis, Cancer
10 mM PBS with 0.05% BSA
8915
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit