Learn More
Bcl-2α Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation, and flow cytometry experiments.
Supplier: Epredia MS123P1ABX
Description
Expression of Bcl-2α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of Bcl-2α protein, whereas the normal or hyperplastic germinal centers are negative.Host Species: Mouse
Clone: 100/D5
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with mouse and rat. Others not known.
Epitope: aa 41-54
Immunogen: A synthetic peptide, aa 41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein
Molecular Weight: 25-26kDa
Positive Control: Raji Cells, Tonsil or follicular lymphomas
Cellular Localization: Cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Denatured verified)
- Flow Cytometry
Specifications
Bcl-2alpha Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Apoptosis | |
Human | |
IgG2a κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
100/D5 | |
Unconjugated | |
A synthetic peptide, aa 41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein | |
IVD | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.