Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Bcl-2α Ab-1, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation, and flow cytometry experiments.

$144.00 - $568.00

Specifications

Antigen Bcl-2alpha Ab-1
Clone 100/D5
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 8
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS123P
View Documents
Thermo Scientific™
MS123P
Purified Ab with BSA and Azide 1mL Each for $526.50
Add to cart
 
MS123P0
View Documents
Thermo Scientific™
MS123P0
Purified Ab with BSA and Azide 100μL Each for $144.00
Add to cart
 
MS123P1
View Documents
Thermo Scientific™
MS123P1
Purified Ab with BSA and Azide 500μL Each for $391.00
Add to cart
 
MS123P1ABX
View Documents
Thermo Scientific™
MS123P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $443.50
Add to cart
 
MS123PABX
View Documents
Thermo Scientific™
MS123PABX
Ab without BSA/Azide (Frozen) 200μL Each for $568.00
Add to cart
 
MS123R7
View Documents
Thermo Scientific™
MS123R7
Ready-to-use for IHC (UVLP) 7mL Each for $279.50
Add to cart
 
MS123RQ
View Documents
Thermo Scientific™
MS123RQ
Ready-to-use for IHC (Quanto) 12mL Each for $414.00
Add to cart
 
MS123R1
View Documents
Thermo Scientific™
MS123R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence, immunoprecipitation, and flow cytometry experiments with Thermo Scientific™ Bcl-2α Ab-1, Mouse Monoclonal Antibody.

Expression of Bcl-2α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of Bcl-2α protein, whereas the normal or hyperplastic germinal centers are negative.

Host Species: Mouse

Clone: 100/D5

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: aa 41-54

Immunogen: A synthetic peptide, aa 41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein

Molecular Weight: 25-26kDa

Positive Control: Raji Cells, Tonsil or follicular lymphomas

Cellular Localization: Cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Denatured verified)
  • Flow Cytometry

Specifications

Specifications

Bcl-2alpha Ab-1
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
IVD
Primary
100/D5
Monoclonal
Murine
Apoptosis
A synthetic peptide, aa 41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit