Learn More
Bcl-2 Antibody (OTI9D3), DyLight 680, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270245FR
Catalog No. NBP270245FR
Add to Cart
Description
Description
Bcl-2 Monoclonal specifically detects Bcl-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
Bcl-2
Monoclonal
DyLight 680
PBS with 0.05% Sodium Azide
BCL2
Full length recombinant protein of human Bcl-2 (NP_000624) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG2b
Western Blot, Immunohistochemistry
OTI9D3
Western Blot, Immunohistochemistry
apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2
Mouse
Affinity Purified
Apoptosis, Autophagy, Biologically Active Proteins, Cancer, Cellular Markers, Mitochondrial Mediated Pathway, Mitophagy, Oncogenes, Phospho Specific, Tumor Suppressors
596.0
Store at 4°C in the dark.
Recommendations
Recommendations