Learn More
Enzo Life SciencesSupplier Diversity Partner Bcl-2 (human), ELISA kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI900133
The Bcl-2 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours that measures pan Bcl-2. Absorbance is read at 450 nm. Obtain results from up to 39 samples in duplicate per 96-well kit. The Bcl-2 (human), EIA kit provides fully quantitative results that surpass semi-quantitative Western blot analysis and easy-to-use liquid color-coded reagents that reduce error. Sensitivity: 3.8 pg/ml (range 18.8 - 1,200 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human. Contents: Microtiter plate, Conjugate, Antibody, Assay buffer 13, Wash buffer concentrate, Standard, TMB Substrate, Cell lysis buffer 2, Stop solution 2
Catalog No. 50-658-164