Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 Mouse anti-Human, FITC, Clone: OTI9D3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270245F

Catalog No. NBP270245F


Add to cart

Description

Description

Bcl-2 Monoclonal antibody specifically detects Bcl-2 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Bcl-2
OTI9D3
Immunohistochemistry
Affinity Purified, Conjugated
BCL2
Full length recombinant protein of human Bcl-2 (NP_000624) produced in HEK293T cells.
Immunogen affinity purified
Apoptosis, Autophagy, Biologically Active Proteins, Cancer, Cellular Markers, Mitochondrial Mediated Pathway, Mitophagy, Oncogenes, Phospho Specific, Tumor Suppressors
Primary
596.0
Immunohistochemistry
FITC
PBS with 0.05% Sodium Azide
apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit