Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-2 Mouse, Clone: 100/D5+124, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2344440.02MG
Description
Bcl-2 Monoclonal specifically detects Bcl-2 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Bcl-2 | |
Monoclonal | |
Unconjugated | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2 | |
Mouse | |
25 kDa | |
0.02 mg | |
Apoptosis, Autophagy, Cancer, Cellular Markers, Tumor Suppressors | |
596 | |
Human, Mouse (Negative), Rat (Negative) | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
100/D5+124 | |
Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/mlimmunohistochemistry-Paraffin 0.5-1ug/ml, SDS-Page | |
P10415 | |
BCL2 | |
A synthetic peptide, aa 41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 protein. | |
Protein A purified | |
RUO | |
Primary | |
This antibody recognizes a protein of 25-26kDa, identified as the bcl-2 alpha oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Expression of bcl-2 alpha oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of bcl-2 alpha protein, whereas the normal or hyperplastic germinal centers are negative. Consequently, this antibody is valuable when distinguishing between reactive and neoplastic follicular proliferation in lymph node biopsies. It may also be used in distinguishing between those follicular lymphomas that express bcl-2 protein and the small number in which the neoplastic cells are bcl-2 negative. | |
Store at 4C. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title