Learn More
Bcl-2 related protein A1 Antibody (OTI3D10), HRP, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270246H
Catalog No. NBP270246H
Add to Cart
Description
Description
Bcl-2 related protein A1 Monoclonal specifically detects Bcl-2 related protein A1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
Bcl-2 related protein A1
Monoclonal
HRP
PBS with No Preservative
BCL2A1
Full length recombinant protein of human Bcl-2 related protein A1 (NP_004040) produced in E.coli.
0.1 mL
Primary
Human
IgG1
Western Blot
OTI3D10
Western Blot
ACC-1, ACC-2, Bcl2-L-5, BCL2L5HBPA1, Bcl-2-like protein 5, BCL2-related protein A1, BFL1bcl-2-related protein A1, GRSbcl2-L-5, hematopoietic BCL2-related protein A1, Hemopoietic-specific early response protein, Protein BFL-1, Protein GRS
Mouse
Affinity Purified
Apoptosis, Cancer
597.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions