Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 related protein A1 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI3D10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270246AF700

Catalog No. N270246A700


Add to cart

Description

Description

Bcl-2 related protein A1 Monoclonal antibody specifically detects Bcl-2 related protein A1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Bcl-2 related protein A1
OTI3D10
Western Blot
Affinity Purified, Conjugated
BCL2A1
Full length recombinant protein of human Bcl-2 related protein A1 (NP_004040) produced in E.coli.
Immunogen affinity purified
Apoptosis, Cancer
Primary
597.0
Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
ACC-1, ACC-2, Bcl2-L-5, BCL2L5HBPA1, Bcl-2-like protein 5, BCL2-related protein A1, BFL1bcl-2-related protein A1, GRSbcl2-L-5, hematopoietic BCL2-related protein A1, Hemopoietic-specific early response protein, Protein BFL-1, Protein GRS
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit