Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-3 Polyclonal Antibody
Bcl-3 Polyclonal Antibody
Supplier: Thermo Scientific PA5116531
Description
Bcl-3 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
Bcl-3 | |
Polyclonal | |
Unconjugated | |
Bcl3 | |
AI528691; B cell leukemia/lymphoma 3; B-cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein; B-cell lymphoma 3 protein homolog; B-cell lymphoma 3-encoded protein; BCL3; BCL-3; Bcl-3 protein; BCL4; chronic lymphatic leukemia protein; D19S37; Proto-oncogene BCL3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12051, 602, 680611 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
4.09 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P20749, Q9Z2F6 | |
Bcl3 | |
A synthetic peptide corresponding to a sequence within amino acids 300-400 of human BCL3 (NP_0051692). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction