Learn More
Bcl-6 Mouse anti-Human, Clone: BCL6/1475, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
$240.50 - $482.00
Specifications
Antigen | Bcl-6 |
---|---|
Clone | BCL6/1475 |
Concentration | 0.2 mg/mL |
Dilution | Western Blot 0.5-1 ug/ml, Flow Cytometry 0.5-1 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, SDS-Page, Protein Array 1:100-1:2000 |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25959620
|
Novus Biologicals
NBP25959620UG |
20 μg |
Each for $240.50
|
|
|||||
NBP259596X
|
Novus Biologicals
NBP259596100UG |
100 μg |
Each for $482.00
|
|
|||||
Description
Bcl-6 Monoclonal specifically detects Bcl-6 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein Array.Specifications
Bcl-6 | |
0.2 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, SDS-Page | |
Unconjugated | |
Mouse | |
Cancer, Cell Cycle and Replication, Tumor Suppressors | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
604 | |
Recombinant human bcl-6 protein fragment (aa256-389) | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
95 kDa |
BCL6/1475 | |
Western Blot 0.5-1 ug/ml, Flow Cytometry 0.5-1 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, SDS-Page, Protein Array 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
B-cell CLL/lymphoma 6, B-cell lymphoma 6 protein transcript, BCL-5, BCL5B-cell lymphoma 5 protein, BCL-6, BCL6A, cys-his2 zinc finger transcription factor, LAZ3ZBTB27B-cell lymphoma 6 protein, lymphoma-associated zinc finger gene on chromosome 3, Protein LAZ-3, Zinc finger and BTB domain-containing protein 27, Zinc finger protein 51ZNF51, zinc finger transcription factor BCL6S | |
BCL6 | |
IgG1 κ | |
Protein A or G purified | |
The specificity of this monoclonal antibody to its intended target was validated by HuProt(TM) Array, containing more than 19,000, full-length human proteins. Recognizes a protein of 95kDa, which is identified as Bcl-6. Antibody to bcl-6 is helpful in a number of diagnostic settings: (1) In the differential diagnosis of small B-cell lymphoma. Follicular lymphoma will show bcl-6 (and CD10) positivity whereas other small B-cell lymphomas are usually negative. (2) Bcl-6 is an important prognostic marker in diffuse large B-cell lymphomas (DLBCL), where CD10, bcl-6 and MUM1/IRF4 are used to identify germinal center and activated B-cell phenotypes. (3) Bcl-6 can be valuable in distinguishing classical Hodgkin lymphoma from nodular lymphocyte predominant Hodgkin lymphoma (NLPHL). The Reed-Sternberg cells of classical Hodgkin lymphoma are bcl-6 negative whereas the large (L&H) cells of NLPHL are bcl-6 positive. In contrast, anti-Bcl-6 rarely stains mantle-cell lymphoma and MALT lymphoma |
For Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.