Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-6 Mouse anti-Human, Clone: BCL6/1526, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30752710UG

 View more versions of this product

Catalog No. NB122527


Add to cart

Description

Description

Bcl-6 Monoclonal antibody specifically detects Bcl-6 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

Bcl-6
BCL6/1526
Unconjugated
10 mM PBS with 0.05% BSA
B-cell CLL/lymphoma 6, B-cell lymphoma 6 protein transcript, BCL-5, BCL5B-cell lymphoma 5 protein, BCL-6, BCL6A, cys-his2 zinc finger transcription factor, LAZ3ZBTB27B-cell lymphoma 6 protein, lymphoma-associated zinc finger gene on chromosome 3, Protein LAZ-3, Zinc finger and BTB domain-containing protein 27, Zinc finger protein 51ZNF51, zinc finger transcription factor BCL6S
Recombinant human Bcl-6 protein fragment (aa256-389) (Exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Immunohistochemistry (Paraffin), Peptide Array
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Purified
Mouse
IgG1 Kappa
100 μg
Cancer, Cell Cycle and Replication, Tumor Suppressors
Primary
604
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit