Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL6 Mouse anti-Human, Mouse, Clone: GI191E, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 14988780

 View more versions of this product

Catalog No. 50-138-24


Add to cart

Description

Description

Description: This GI191E monoclonal antibody reacts with mouse and human Bcl-6, which is also known as B-cell lymphoma 6. A member of the POZ family of transcriptional repressors, Bcl-6 is expressed most abundantly in germinal center B cells, as well as in differentiating and activated B cells. This 79-kDa protein is essential for germinal center B cell proliferation and survival. Moreover, during B cell development, Bcl-6 plays an important role in promoting pre-B cell survival to enable the generation of a diverse B cell repertoire. Moreover, Bcl-6 is expressed constitutively in diffuse large B cell lymphomas (DLBCLs), Burkitt ft.s lymphoma, and non-Hodgkin ft.s lymphoma. Reports also suggest expression of Bcl-6 in CD4+ T follicular helper cells and memory T cells. Bcl-6 has been reported to be antagonized by Blimp-1. Applications Reported: This GI191E antibody has been reported for use in immunoblotting (WB) and immunohistochemistry (FFPE). Applications Tested: This GI191E antibody has been tested by immunoblotting of reduced cell lysate prepared from A20 cells and by immunohistochemistry on human tissue using high pH antigen retrieval. This antibody can be used at less than or equal to 5 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.

2 µm post-manufacturing filtered. Bcl-6 proto-oncogene product (a Kruppel-type zinc-finger protein) is mainly expressed in normal germinal center B cells and related lymphomas. Bcl-6 is involved in chromosome rearrangements at 3q27 in non-Hodgkin ft.s lymphomas and Bcl-6 rearrangements have been detected in 33%-45% of diffuse large B cell lymphomas. Bcl-6 has been detected immunohistochemically in follicular lymphomas, diffuse large B cell lymphomas, Burkitt ft.s lymphomas and in nodular, lymphocyte predomit Hodgkin ft.s disease.
Specifications

Specifications

BCL6
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P41182, P41183
B-cell lymphoma 6 protein; BCL-6; Zinc finger protein 51; LAZ-3 protein; BCL-5; Zinc finger and BTB domain-containing protein 27; ZNF51; ZBTB27; LAZ3; BCL5; BCL6
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
GI191E
Unconjugated
BCL6
Liquid
Bcl6
IgG1, kappa
25 μg
4° C
Primary
12053, 604
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.