Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL-6 Mouse anti-Mouse,Human,Rat, Unconjugated, Clone: GI191E/A8, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen BCL-6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABT13181MI
View Documents
MilliporeSigma
MABT1318100UG
100 μg
Each for $386.00
Add to cart
 
MABT13182MI
View Documents
MilliporeSigma
MABT131825UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

BCL-6 Monoclonal antibody specifically detects BCL-6 in Mouse,Human,Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

BCL-6
Unconjugated
Mouse
B-cell lymphoma 6 protein;B-cell lymphoma 5 protein;BCL-5;Protein LAZ-3;Zinc finger and BTB domain-containing protein 27;Zinc finger protein 51
pCDNA3‐BCL6 plasmid vector (full length sequence of human BCL-6).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
ELISA, Immunohistochemistry (Paraffin), Western Blot
Purified
Mouse, Human, Rat
BCL6, BCL5, LAZ3, ZBTB27, ZNF51
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit