Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL6 Rat anti-Mouse, PerCP-eFluor™ 710, Clone: BCL-DWN, eBioscience™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen 46545380

 View more versions of this product

Catalog No. 50-112-3827


Add to cart

Description

Description

Description: This BCL-DWN monoclonal antibody reacts with mouse Bcl-6, which is also known as B-cell lymphoma 6. A member of the POZ family of transcriptional repressors, Bcl-6 is expressed most abundantly in germinal center B cells, as well as in differentiating and activated B cells. This 79-kDa protein is essential for germinal center B cell proliferation and survival. Moreover, during B cell development, Bcl-6 plays an important role in promoting pre-B cell survival to enable the generation of a diverse B cell repertoire. Moreover, Bcl-6 is expressed constitutively in diffuse large B cell lymphomas (DLBCLs), Burkitt ft.s lymphoma, and non-Hodgkin ft.s lymphoma. Reports also suggest expression of Bcl-6 in CD4+ T follicular helper cells and memory T cells. Bcl-6 has been reported to be antagonized by Blimp-1. Applications Reported: This BCL-DWN antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This BCL-DWN antibody has been tested by flow cytometric analysis of splenocytes isolated from sheep red blood cell-immunized mice. This can be used at less than or equal to 0.015 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 µm post-manufacturing filtered. Bcl-6 proto-oncogene product (a Kruppel-type zinc-finger protein) is mainly expressed in normal germinal center B cells and related lymphomas. Bcl-6 is involved in chromosome rearrangements at 3q27 in non-Hodgkin ft.s lymphomas and Bcl-6 rearrangements have been detected in 33%-45% of diffuse large B cell lymphomas. Bcl-6 has been detected immunohistochemically in follicular lymphomas, diffuse large B cell lymphomas, Burkitt ft.s lymphomas and in nodular, lymphocyte predomit Hodgkin ft.s disease.
Specifications

Specifications

BCL6
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P41183
B-cell lymphoma 6 protein; BCL-6; Zinc finger protein 51; LAZ-3 protein; BCL-5; Zinc finger and BTB domain-containing protein 27; ZNF51; ZBTB27; LAZ3; BCL5; BCL6
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
BCL-DWN
PerCP-eFluor™ 710
Bcl6
Liquid
Bcl6
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12053
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.