Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Bcl-G Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA14210

Catalog No. PA14210


Only null left
Add to Cart

Description

Description

PA1-4210 detects the long form of BCL-G in human samples. PA1-4210 has been successfully used in Western blot procedures. The PA1-4210 immunogen is a synthetic peptide corresponding to residues S(71) W P C R N S Q S S E K A(83) of the long form of human Bcl-G.

The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. Overexpression of this gene has been shown to induce apoptosis in cells. Three alternatively spliced transcript variants encoding two distinct isoforms have been reported for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bcl-G
Polyclonal
Unconjugated
BCL2L14
4930452K23Rik; 4933405K19Rik; 9030625M01Rik; AI429211; Apoptosis facilitator Bcl-2-like protein 14; apoptosis regulator BCL-G; BCL2 like 14; Bcl2l14; bcl2-L-14; BCL2-like 14 (apoptosis facilitator); BCLG; testicular tissue protein Li 26
Rabbit
Affinity chromatography
RUO
79370
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9BZR8
BCL2L14
Synthetic peptide corresponding to residues S(71) W P C R N S Q S S E K A(83) of the long form of human Bcl-G.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.