Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Bcl-X Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific™ Bcl-X Ab-1, Mouse Monoclonal Antibody.

Specifications

Antigen Bcl-X Ab-1
Clone 2H12
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Host Species Murine
View More Specs

Products 2
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS715R1
View Documents
Thermo Scientific™
MS715R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS715P0
View Documents
Thermo Scientific™
MS715P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments with Thermo Scientific™ Bcl-X Ab-1, Mouse Monoclonal Antibody.

Bcl-X has two isoforms, Bcl-XL (long) a 241 amino acid protein and Bcl-XS (short) a 178 amino acid protein lacking a 63 amino acid domain that is well conserved among members of the Bcl-2 family.

Host Species: Mouse

Clone: 2H12

Isotype: IgG2a

Species Reactivity: Human, Mouse, Rat and Pig. Others not tested.

Epitope: aa 3-14

Immunogen: A synthetic peptide, aa 3-14 (Cys-QSNRELVVDFLS) of human Bcl-X protein. This amino acid sequence is shared by human and murine Bcl-X protein.

Molecular Weight: 27kDa

Positive Control: Hodgkin's lymphoma

Cellular Localization: Cytoplasmic and Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not optimum)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Bcl-X Ab-1
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Murine
Apoptosis
A synthetic peptide, aa 3-14 (Cys-QSNRELVVDFLS) of human Bcl-X protein. This amino acid sequence is shared by human and murine bcl-X protein.
2H12
Monoclonal
RUO
Primary
IgG2a
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit